siRNA Details
| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012914.1 | NP_037046.1 | 11 | 1432 | ||
| XM_006246597.2 | XP_006246659.1 | 12 | 2046 | |||
| GenBank mRNA | M30581.1 | - | 11 | 1432 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2a3| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012914.1 | NP_037046.1 | 11 | 1432 | ||
| XM_006246597.2 | XP_006246659.1 | 12 | 2046 | |||
| GenBank mRNA | M30581.1 | - | 11 | 1432 | ||