siRNA Details
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012931.1 | NP_037063.1 | 1 | 282 | ||
GenBank mRNA | D29766.1 | - | 1 | 282 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012931.1 | NP_037063.1 | 1 | 282 | ||
GenBank mRNA | D29766.1 | - | 1 | 282 |