siRNA Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 3 | 514 | ||
| XM_006251084.2 | XP_006251146.1 | 3 | 748 | |||
| GenBank mRNA | FM031363.1 | - | 3 | 514 | ||
| U52095.1 | - | 3 | 308 | |||
| U52102.1 | - | 3 | 482 | |||

Crmp1
Genomic Map