siRNA Details
| Gene Symbol: Cyp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012940.2 | NP_037072.1 | 2 | 1333 | ||
| GenBank mRNA | U09540.1 | - | 1333 | |||
| X78583.1 | - | 1 | 23 | |||
| X83867.1 | - | 2 | 1334 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp1b1| Gene Symbol: Cyp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012940.2 | NP_037072.1 | 2 | 1333 | ||
| GenBank mRNA | U09540.1 | - | 1333 | |||
| X78583.1 | - | 1 | 23 | |||
| X83867.1 | - | 2 | 1334 | |||