siRNA Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012955.1 | NP_037087.1 | 3 | 306 | ||
| XM_006257215.2 | XP_006257277.1 | 3 | 444 | |||
| XM_006257216.2 | XP_006257278.1 | 3 | 605 | |||
| XM_006257217.2 | XP_006257279.1 | 2 | 319 | |||
| XM_008773403.1 | XP_008771625.1 | 2 | 660 | |||
| GenBank mRNA | X90355.1 | - | 3 | 306 | ||

Cenpi
Genomic Map