siRNA Details
| Gene Symbol: Ndufa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012985.2 | NP_037117.1 | 2, 3 | 221 | ||
| GenBank mRNA | BC059148.1 | - | 2, 3 | 84 | 6919202 | |
| D86215.1 | - | 2, 3 | 221 | |||
| FQ212236.1 | - | 2, 3 | 82 | |||
| FQ228773.1 | - | 2, 3 | 82 | |||
| FQ229037.1 | - | 2, 3 | 81 | |||

Ndufa5
Genomic Map