siRNA Details
| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ493658.1 | - | 4 | 259 | ||
| BC059147.1 | - | 3 | 139 | 6888437 | ||
| BG665749.1 | - | 3 | 138 | |||
| BG666168.1 | - | 3 | 129 | |||
| CB613293.1 | - | 3 | 134 | |||
| M24852.1 | - | 3 | 126 | |||

Pcp4
Genomic Map