siRNA Details
Gene Symbol: Ptgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013015.2 | NP_037147.1 | 2, 3 | 296 | ||
GenBank mRNA | FQ214359.1 | - | 1, 2 | 548 | ||
J04488.1 | - | 2, 3 | 296 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013015.2 | NP_037147.1 | 2, 3 | 296 | ||
GenBank mRNA | FQ214359.1 | - | 1, 2 | 548 | ||
J04488.1 | - | 2, 3 | 296 |