siRNA Details
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767547.1 | XP_008765769.1 | 1 | 105 | ||
GenBank mRNA | J02675.1 | - | 1 | 129 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767547.1 | XP_008765769.1 | 1 | 105 | ||
GenBank mRNA | J02675.1 | - | 1 | 129 |