siRNA Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013028.1 | NP_037160.1 | 2 | 142 | ||
| GenBank mRNA | AJ002258.1 | - | 2 | 163 | ||
| AJ002259.1 | - | 2 | 142 | |||
| AJ002260.1 | - | 1 | 115 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013665, U67055, AK032007, AK045937, U66918, and others.

Shox2
Genomic Map