siRNA Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 4 | 678 | ||
| XM_006251084.2 | XP_006251146.1 | 4 | 912 | |||
| GenBank mRNA | U52095.1 | - | 4 | 472 | ||
| U52102.1 | - | 4 | 646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crmp1| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 4 | 678 | ||
| XM_006251084.2 | XP_006251146.1 | 4 | 912 | |||
| GenBank mRNA | U52095.1 | - | 4 | 472 | ||
| U52102.1 | - | 4 | 646 | |||