siRNA Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012949.2 | NP_037081.2 | 7 | 557 | ||
| XM_006246599.2 | XP_006246661.1 | 7 | 628 | |||
| XM_006246600.2 | XP_006246662.1 | 7 | 608 | |||
| GenBank mRNA | BC083566.1 | - | 7 | 544 | 7193009 | |
| DY321069.1 | - | 7 | 557 | |||
| FQ214810.1 | - | 6 | 535 | |||
| FQ214945.1 | - | 7 | 575 | |||
| FQ214984.1 | - | 7 | 567 | |||
| FQ215034.1 | - | 3 | 234 | |||
| FQ215197.1 | - | 3 | 234 | |||
| FQ215221.1 | - | 6 | 470 | |||
| FQ215310.1 | - | 3 | 234 | |||
| FQ215357.1 | - | 7 | 579 | |||
| FQ215526.1 | - | 7 | 567 | |||
| FQ215607.1 | - | 3 | 234 | |||
| FQ215648.1 | - | 7 | 567 | |||
| FQ215759.1 | - | 7 | 579 | |||
| FQ215780.1 | - | 3 | 234 | |||
| FQ215843.1 | - | 6 | 534 | |||
| FQ215865.1 | - | 7 | 567 | |||
| FQ216020.1 | - | 3 | 234 | |||
| FQ216150.1 | - | 6 | 535 | |||
| FQ216271.1 | - | 3 | 234 | |||
| FQ216382.1 | - | 7 | 567 | |||
| FQ216386.1 | - | 6 | 544 | |||
| FQ216660.1 | - | 6 | 534 | |||
| FQ216781.1 | - | 3 | 234 | |||
| FQ217095.1 | - | 7 | 567 | |||
| FQ223831.1 | - | 3 | 234 | |||
| FQ224938.1 | - | 7 | 567 | |||
| Y00979.1 | - | 6 | 519 | |||

Eno3
Genomic Map