siRNA Details
| Gene Symbol: Mmp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012980.2 | NP_037112.2 | 5 | 854 | ||
| GenBank mRNA | BC099781.1 | - | 5 | 848 | 7933545 | |
| CK602095.1 | - | 1 | 103 | |||
| U46034.1 | - | 5 | 854 | |||
| Gene Symbol: LOC103694874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773828.1 | XP_008772050.1 | 5 | 880 | ||

Mmp11
Genomic Map