siRNA Details
| Gene Symbol: Psap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190236.1 | NP_001177165.1 | 4 | 406 | ||
| NM_001190237.1 | NP_001177166.1 | 4 | 406 | |||
| NM_001190238.1 | NP_001177167.1 | 4 | 406 | |||
| NM_013013.2 | NP_037145.2 | 4 | 406 | |||
| XM_006256403.1 | XP_006256465.1 | 4 | 418 | |||
| GenBank mRNA | BC061759.1 | - | 4 | 340 | 5600088 | |
| CK365248.1 | - | 4 | 406 | |||
| CO403947.1 | - | 2 | 174 | |||
| FQ217536.1 | - | 4 | 357 | |||
| M19936.1 | - | 4 | 356 | |||
| S81353.1 | - | 4 | 356 | |||

Psap
Genomic Map