siRNA Details
| Gene Symbol: Pspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013014.1 | NP_037146.1 | 2 | 263 | ||
| GenBank mRNA | AF040961.1 | - | 2 | 263 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pspn| Gene Symbol: Pspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013014.1 | NP_037146.1 | 2 | 263 | ||
| GenBank mRNA | AF040961.1 | - | 2 | 263 | ||