siRNA Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013019.2 | NP_037151.2 | 5 | 558 | ||
| XM_008772599.1 | XP_008770821.1 | 5 | 1397 | |||
| XM_008772600.1 | XP_008770822.1 | 5 | 571 | |||
| GenBank mRNA | BC062016.1 | - | 5 | 558 | 5621494 | |
| X06890.1 | - | 5 | 490 | |||

Rab4a
Genomic Map