siRNA Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013028.1 | NP_037160.1 | 2 | 212 | ||
| GenBank mRNA | AJ002258.1 | - | 2 | 233 | ||
| AJ002259.1 | - | 2 | 212 | |||
| AJ002260.1 | - | 1 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shox2| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013028.1 | NP_037160.1 | 2 | 212 | ||
| GenBank mRNA | AJ002258.1 | - | 2 | 233 | ||
| AJ002259.1 | - | 2 | 212 | |||
| AJ002260.1 | - | 1 | 185 | |||