siRNA Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013031.1 | NP_037163.1 | 11, 12 | 1181 | ||
| XM_006231664.2 | XP_006231726.1 | 11, 12 | 2022 | |||
| XM_008760515.1 | XP_008758737.1 | 10, 11 | 1381 | |||
| XM_008760516.1 | XP_008758738.1 | 11, 12 | 2027 | |||
| XM_008760517.1 | XP_008758739.1 | 11, 12 | 2022 | |||
| XM_008760518.1 | XP_008758740.1 | 10, 11 | 1381 | |||
| XM_008760519.1 | XP_008758741.1 | 9, 10 | 1288 | |||
| XM_008760520.1 | XP_008758742.1 | 8, 9 | 1204 | |||
| GenBank mRNA | L00603.1 | - | 11, 12 | 1181 | ||
| M97381.1 | - | 11, 12 | 1152 | |||

Slc18a2
Genomic Map