siRNA Details
| Gene Symbol: Cnga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012928.1 | NP_037060.1 | 719 | |||
| GenBank mRNA | AF126808.1 | - | 613 | |||
| X55519.1 | - | 719 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnga2| Gene Symbol: Cnga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012928.1 | NP_037060.1 | 719 | |||
| GenBank mRNA | AF126808.1 | - | 613 | |||
| X55519.1 | - | 719 | ||||