siRNA Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 10 | 1254 | ||
| XM_006251084.2 | XP_006251146.1 | 10 | 1488 | |||
| GenBank mRNA | U52095.1 | - | 10 | 1048 | ||
| U52102.1 | - | 10 | 1222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crmp1| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 10 | 1254 | ||
| XM_006251084.2 | XP_006251146.1 | 10 | 1488 | |||
| GenBank mRNA | U52095.1 | - | 10 | 1048 | ||
| U52102.1 | - | 10 | 1222 | |||