siRNA Details
| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012937.1 | NP_037069.1 | 4 | 349 | ||
| GenBank mRNA | X16072.1 | - | 4 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crybb2| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012937.1 | NP_037069.1 | 4 | 349 | ||
| GenBank mRNA | X16072.1 | - | 4 | 349 | ||