siRNA Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270942.1 | NP_001257871.1 | 3 | 414 | ||
| NM_001270943.1 | NP_001257872.1 | 2 | 181 | |||
| NM_001270944.1 | NP_001257873.1 | 2 | 145 | |||
| NM_003709.3 | NP_003700.1 | 2 | 624 | |||
| XM_005246926.3 | XP_005246983.1 | 3 | 410 | |||
| XM_006712815.3 | XP_006712878.1 | 3 | 414 | |||
| XM_011512072.2 | XP_011510374.1 | 3 | 588 | |||
| XM_011512073.2 | XP_011510375.1 | 4 | 596 | |||
| XM_011512074.2 | XP_011510376.1 | 3 | 191 | |||
| XM_011512075.1 | XP_011510377.1 | 2 | 597 | |||
| XM_017005161.1 | XP_016860650.1 | 3 | 410 | |||
| XM_017005162.1 | XP_016860651.1 | 2 | 160 | |||
| XM_017005163.1 | XP_016860652.1 | 2 | 597 | |||
| XM_017005164.1 | XP_016860653.1 | 2 | 609 | |||
| GenBank mRNA | AB015132.1 | - | 2 | 499 | ||
| AK027371.1 | - | 2 | 325 | |||
| AK297312.1 | - | 2 | 132 | |||
| AK314439.1 | - | 2 | 555 | |||
| BC012919.1 | - | 3 | 405 | 4509200 | ||
| BX537890.1 | - | 2 | 181 | |||
| CR542112.1 | - | 2 | 176 | |||
| DA800775.1 | - | 3 | 414 | |||
| DC370575.1 | - | 2 | 145 | |||

KLF7
Genomic Map