siRNA Details
| Gene Symbol: Emd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012948.1 | NP_037080.1 | 4, 5 | 525 | ||
| GenBank mRNA | BC059151.1 | - | 4, 5 | 538 | 6919042 | |
| X98377.1 | - | 4, 5 | 525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emd| Gene Symbol: Emd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012948.1 | NP_037080.1 | 4, 5 | 525 | ||
| GenBank mRNA | BC059151.1 | - | 4, 5 | 538 | 6919042 | |
| X98377.1 | - | 4, 5 | 525 | |||