siRNA Details
Gene Symbol: Gpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012961.1 | NP_037093.1 | 1 | 965 | ||
XM_008767077.1 | XP_008765299.1 | 2 | 1159 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
