siRNA Details
| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012962.1 | NP_037094.1 | 7 | 691 | ||
| XM_008762303.1 | XP_008760525.1 | 7 | 1105 | |||
| GenBank mRNA | BC078700.1 | - | 6 | 656 | 7110971 | |
| L38615.1 | - | 7 | 691 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gss| Gene Symbol: Gss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012962.1 | NP_037094.1 | 7 | 691 | ||
| XM_008762303.1 | XP_008760525.1 | 7 | 1105 | |||
| GenBank mRNA | BC078700.1 | - | 6 | 656 | 7110971 | |
| L38615.1 | - | 7 | 691 | |||