siRNA Details
| Gene Symbol: Mras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012981.2 | NP_037113.2 | 3 | 449 | ||
| GenBank mRNA | BC085752.1 | - | 3 | 449 | 7130510 | |
| D89863.1 | - | 857 | ||||
| Gene Symbol: Lmntd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191980.1 | NP_001178909.1 | 8, 9 | 1212 | ||
| XM_008763419.1 | XP_008761641.1 | 8, 9 | 2555 | |||
| XM_008763420.1 | XP_008761642.1 | 7, 8 | 1120 | |||
| XM_008763421.1 | XP_008761643.1 | 6, 7 | 1164 | |||

Mras
Genomic Map