siRNA Details
| Gene Symbol: Nes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012987.1 | NP_037119.1 | 5 | 3362 | ||
| XM_006232600.2 | XP_006232662.1 | 4 | 3944 | |||
| GenBank mRNA | AF538924.1 | - | 4 | 3499 | ||
| M34384.1 | - | 5 | 3362 | |||
| Gene Symbol: LOC100910255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232557.2 | XP_006232619.1 | 4 | 3945 | ||

Nes
Genomic Map