siRNA Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013016.2 | NP_037148.2 | 5 | 1161 | ||
| XM_006234951.2 | XP_006235013.1 | 4 | 1459 | |||
| XM_006234952.2 | XP_006235014.1 | 5 | 1821 | |||
| XM_006234953.2 | XP_006235015.1 | 5 | 2003 | |||
| GenBank mRNA | AF055065.1 | - | 5 | 1025 | ||
| BC099773.1 | - | 5 | 1097 | 7932298 | ||
| D38468.1 | - | 4 | 1216 | |||
| D85183.1 | - | 4 | 973 | |||
| U62328.1 | - | 5 | 1161 | |||

Sirpa
Genomic Map