siRNA Details
| Gene Symbol: Rock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013022.2 | NP_037154.2 | 31 | 3834 | ||
| XM_006239909.2 | XP_006239971.1 | 31 | 3864 | |||
| XM_008764574.1 | XP_008762796.1 | 32 | 4035 | |||
| XM_008764575.1 | XP_008762797.1 | 32 | 4035 | |||
| GenBank mRNA | U38481.1 | - | 31 | 3832 | ||

Rock2
Genomic Map