siRNA Details
| Gene Symbol: Slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013032.3 | NP_037164.3 | 10 | 1152 | ||
| GenBank mRNA | AF038571.1 | - | 3 | 265 | ||
| BC061743.1 | - | 10 | 1152 | 5599027 | ||
| D63772.1 | - | 1183 | ||||
| L35558.1 | - | 5 | 598 | |||
| U21104.1 | - | 10 | 1164 | |||
| U39555.1 | - | 10 | 1242 | |||
| X94255.1 | - | 10 | 1209 | |||

Slc1a1
Genomic Map