siRNA Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013055.1 | NP_037187.1 | 4 | 578 | ||
| XM_006242381.2 | XP_006242443.1 | 4 | 1959 | |||
| XM_006242382.2 | XP_006242444.1 | 2 | 192 | |||
| XM_008765675.1 | XP_008763897.1 | 1 | 922 | |||
| GenBank mRNA | D49785.1 | - | 4 | 578 | ||
| FQ211695.1 | - | 4 | 428 | |||
| FQ212412.1 | - | 4 | 564 | |||

Map3k12
Genomic Map