siRNA Details
| Gene Symbol: Ptk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013081.2 | NP_037213.1 | 3 | 763 | ||
| XM_008765546.1 | XP_008763768.1 | 5 | 892 | |||
| GenBank mRNA | AF020777.1 | - | 3 | 762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptk2| Gene Symbol: Ptk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013081.2 | NP_037213.1 | 3 | 763 | ||
| XM_008765546.1 | XP_008763768.1 | 5 | 892 | |||
| GenBank mRNA | AF020777.1 | - | 3 | 762 | ||