siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013095.3 | NP_037227.1 | 2 | 561 | ||
| GenBank mRNA | BC064437.1 | - | 561 | 5599910 | ||
| U66479.1 | - | 2 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad3| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013095.3 | NP_037227.1 | 2 | 561 | ||
| GenBank mRNA | BC064437.1 | - | 561 | 5599910 | ||
| U66479.1 | - | 2 | 370 | |||