siRNA Details
| Gene Symbol: Mc4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013099.2 | NP_037231.1 | 1 | 531 | ||
| GenBank mRNA | CB584477.1 | - | 1 | 531 | ||
| U67863.1 | - | 1 | 270 | |||
More Information
Other Species Targeted
Set Membership: |
|

Mc4r
Genomic Map