siRNA Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013111.3 | NP_037243.2 | 807 | |||
| XM_008768943.1 | XP_008767165.1 | 2, 3 | 1079 | |||
| XM_008768944.1 | XP_008767166.1 | 2, 3 | 1079 | |||
| GenBank mRNA | AB066224.1 | - | 2, 3 | 564 | ||
| AF245000.1 | - | 747 | ||||
| D67087.1 | - | 2, 3 | 537 | |||
| L10152.1 | - | 2, 3 | 531 | |||
| U70476.1 | - | 2, 3 | 616 | |||

Slc7a1
Genomic Map