siRNA Details
| Gene Symbol: Parp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013063.2 | NP_037195.1 | 4 | 616 | ||
| GenBank mRNA | BC085765.1 | - | 4 | 616 | 7107130 | |
| U94340.1 | - | 4 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parp1| Gene Symbol: Parp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013063.2 | NP_037195.1 | 4 | 616 | ||
| GenBank mRNA | BC085765.1 | - | 4 | 616 | 7107130 | |
| U94340.1 | - | 4 | 584 | |||