siRNA Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB100593.1 | - | 2 | 272 | ||
| AB100594.1 | - | 2 | 272 | |||
| BC061756.1 | - | 2 | 276 | 5600029 | ||
| X05300.1 | - | 2 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpn1| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB100593.1 | - | 2 | 272 | ||
| AB100594.1 | - | 2 | 272 | |||
| BC061756.1 | - | 2 | 276 | 5600029 | ||
| X05300.1 | - | 2 | 277 | |||