siRNA Details
| Gene Symbol: Hcrtr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013074.1 | NP_037206.1 | 1 | 245 | ||
| XM_008766215.1 | XP_008764437.1 | 1 | 605 | |||
| GenBank mRNA | AF041246.1 | - | 1 | 245 | ||
More Information
Other Species Targeted
Set Membership: |
|

Hcrtr2
Genomic Map