siRNA Details
| Gene Symbol: Ddr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166022.1 | NP_001159494.1 | 4, 5 | 526 | ||
| NM_013137.2 | NP_037269.2 | 4, 5 | 613 | |||
| GenBank mRNA | BC166604.1 | - | 4, 5 | 613 | ||
| CB613563.1 | - | 4, 5 | 526 | |||

Ddr1
Genomic Map