siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013037.1 | NP_037169.1 | 8 | 1180 | ||
| GenBank mRNA | EU929068.1 | - | 4 | 452 | ||
| U04319.1 | - | 8 | 1180 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rl1| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013037.1 | NP_037169.1 | 8 | 1180 | ||
| GenBank mRNA | EU929068.1 | - | 4 | 452 | ||
| U04319.1 | - | 8 | 1180 | |||