siRNA Details
| Gene Symbol: Tmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013044.2 | NP_037176.2 | 6 | 698 | ||
| GenBank mRNA | BC072521.1 | - | 6 | 698 | 7105834 | |
| U59241.1 | - | 5 | 654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmod1| Gene Symbol: Tmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013044.2 | NP_037176.2 | 6 | 698 | ||
| GenBank mRNA | BC072521.1 | - | 6 | 698 | 7105834 | |
| U59241.1 | - | 5 | 654 | |||