siRNA Details
| Gene Symbol: Sdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013082.3 | NP_037214.1 | 2 | 515 | ||
| XM_006241518.2 | XP_006241580.1 | 2 | 727 | |||
| GenBank mRNA | BC070890.1 | - | 2 | 305 | 7098631 | |
| FQ218442.1 | - | 2 | 248 | |||
| FQ219283.1 | - | 2 | 549 | |||
| FQ219510.1 | - | 2 | 547 | |||
| FQ222974.1 | - | 2 | 597 | |||
| M81687.1 | - | 2 | 515 | |||

Sdc2
Genomic Map