siRNA Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013097.2 | NP_037229.1 | 6, 7 | 835 | ||
| XM_006245829.2 | XP_006245891.1 | 7, 8 | 1515 | |||
| GenBank mRNA | AF397150.1 | - | 6, 7 | 835 | ||
| AF397151.1 | - | 7, 8 | 706 | |||
| BC089764.1 | - | 6, 7 | 835 | 7130586 | ||
| X56060.1 | - | 6, 7 | 688 | |||

Dnase1
Genomic Map