siRNA Details
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013145.1 | NP_037277.1 | 3 | 415 | ||
GenBank mRNA | DQ120469.1 | - | 3 | 289 | ||
M17527.1 | - | 3 | 415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013145.1 | NP_037277.1 | 3 | 415 | ||
GenBank mRNA | DQ120469.1 | - | 3 | 289 | ||
M17527.1 | - | 3 | 415 |