siRNA Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013151.2 | NP_037283.2 | 4 | 293 | ||
| XM_006253347.2 | XP_006253409.1 | 4 | 365 | |||
| XM_008771348.1 | XP_008769570.1 | 5 | 423 | |||
| GenBank mRNA | BC061565.1 | - | 4 | 293 | 6920438 | |
| M23697.1 | - | 3 | 217 | |||
| Gene Symbol: LOC100910418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751623.3 | XP_003751671.2 | 4 | 297 | ||

Plat
Genomic Map