siRNA Details
| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013169.1 | NP_037301.1 | 2 | 159 | ||
| GenBank mRNA | X53430.1 | - | 2 | 159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd3d| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013169.1 | NP_037301.1 | 2 | 159 | ||
| GenBank mRNA | X53430.1 | - | 2 | 159 | ||