siRNA Details
| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013174.2 | NP_037306.1 | 1 | 610 | ||
| GenBank mRNA | BC092195.1 | - | 1 | 597 | 7313608 | |
| U03491.1 | - | 1 | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgfb3| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013174.2 | NP_037306.1 | 1 | 610 | ||
| GenBank mRNA | BC092195.1 | - | 1 | 597 | 7313608 | |
| U03491.1 | - | 1 | 610 | |||