siRNA Details
| Gene Symbol: Gnaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013189.2 | NP_037321.1 | 1 | 601 | ||
| XM_008772866.1 | XP_008771088.1 | 2 | 1376 | |||
| XM_008772867.1 | XP_008771089.1 | 3 | 1511 | |||
| GenBank mRNA | J03773.1 | - | 1 | 601 | ||

Gnaz
Genomic Map