siRNA Details
| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013221.2 | NP_037353.2 | 2 | 133 | ||
| XM_006239975.2 | XP_006240037.1 | 2 | 196 | |||
| GenBank mRNA | BC091165.1 | - | 1 | 107 | 7097013 | |
| CB732549.1 | - | 2 | 189 | |||
| FQ223257.1 | - | 2 | 240 | |||
| FQ224288.1 | - | 2 | 124 | |||
| U09551.1 | - | 1 | 128 | |||

Hbp1
Genomic Map