siRNA Details
| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013225.1 | NP_037357.1 | 3 | 285 | ||
| XM_008763183.1 | XP_008761405.1 | 2 | 179 | |||
| GenBank mRNA | D87248.1 | - | 3 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn6| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013225.1 | NP_037357.1 | 3 | 285 | ||
| XM_008763183.1 | XP_008761405.1 | 2 | 179 | |||
| GenBank mRNA | D87248.1 | - | 3 | 285 | ||